57251
Visiprep™ Manifold Cover (flow control valves and gasket included)
for use with standard, 24-port model
Synonym(s):
24 PORT V.M. COVER W/CONTROL VALVES AND GASKET
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12000000
Recommended Products
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with standard, 24-port model
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Visiprep™ manifold cover (flow control valves and gasket included) is a solid phase extraction vacuum manifold, which allows to continuously process up to 24 solid phase extraction (SPE) tubes. It provides an accuracy in flow control through each solid phase extraction tube by rotating the independent, screw-type valves, thus it ensures that the packing will is not dried in some of the tubes while others finish draining. It also helps in preventing the PTFE solvent guide running through each valve from resting on the work surface and becoming contaminated or damaged.
Footnote
Included with Visiprep 24-port Vacuum Manifold (57250-U)
Legal Information
Visiprep is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service