54075-U
LpDNPH Rezorian solid phase extraction (SPE) Cartridge
bed wt. 350 mg, volume 3 mL, pkg of 50 ea
Synonym(s):
pk50 3mL/350mg DNPH Rezorian Tube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
product name
LpDNPH Rezorian Cartridge, bed wt. 350 mg, volume 3 mL, pkg of 50 ea
Agency
ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016
composition
bed wt., 350 mg
2 4-DNPH (High-purity silica gel)
packaging
pkg of 50 ea
technique(s)
active air sampling: suitable
volume
3 mL
application(s)
air monitoring
environmental
industrial hygiene
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
LpDNPH Rezorian Cartridge have polypropylene components and polyethylene frits, making them solvent-compatible. It can be used in-line with peristaltic pumps and low pressure HPLC systems, or with a syringe and a single sample processor.
Rezorian cartridges are made from a low background polypropylene and feature a luer lock end-fitting. The luer lock enables the user to connect the cartridge to a pump tubing or use two cartridges in a series (piggyback), without an adaptor, to monitor breakthough or to increase sampling capacity.
Application
- Measurement and source characteristics of carbonyl compounds in the atmosphere in Kaohsiung city, Taiwan.: This study explores the atmospheric concentration and source characterization of carbonyl compounds, utilizing the LpDNPH Rezorian Cartridge for sampling and analysis (Wang et al., 2010).
- Identification of the oxidation products of the reaction between alpha-pinene and hydroxyl radicals by gas and high-performance liquid chromatography with mass spectrometric detection.: This research identifies the oxidation products from the interaction between alpha-pinene and hydroxyl radicals, demonstrating the versatility of techniques like those enabled by the LpDNPH Rezorian Cartridge for chemical analysis in chromatographic applications (Van den Bergh et al., 2000).
Legal Information
ASTM is a registered trademark of American Society for Testing and Materials
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service