509426
SGE Gas Tight Syringe, Removable Needle
10MDR-GT, volume 10 mL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Synonym(s):
SYRINGE SGE 10ML, GAS TIGHT
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
eCl@ss:
32070500
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
23 ga
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008900
needle L × O.D.
50 mm × 0.63 mm
volume
10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SGE gas tight syringe with removable needle, Model: 10MDR-GT (volume 10 mL, needle size 23 ga (bevel tip), needle L × O.D. 50 mm × 0.63 mm)
- Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
- Autoclavable syringes but the plunger should be removed.
- Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
- Maximum inertness for liquid or gas samples.
- Suitable for manual dispenser/diluter applications.
- High accuracy of dispensed volume.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service