503207
Carbon Dioxide Trap
volume 250 cc, 1/8 in. fitting
Synonym(s):
CO2 TRAP 250CC, 1/8IN FITTINGS
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
EC Number:
UNSPSC Code:
12000000
Recommended Products
volume
250 cc
fitting
1/8 in. fitting
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Sodium hydroxide nonfibrous silicate material is universally accepted for its high absorptive capacity and indicating properties for carbon dioxide. Typically, this material will absorb 20-30% of its weight in carbon dioxide before replacement of the saturated material is required. As carbon dioxide is absorbed, the greenish-brown material turns white because of the formation of sodium carbonate. The trap body is constructed of borosilicate glass, and the fittings are nickel-plated and have sintered stainless steel frits.
A moisture trap should be installed downstream from this unit to absorb the moisture that is produced.
Note: Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent.
Dimensions:
A moisture trap should be installed downstream from this unit to absorb the moisture that is produced.
Note: Exercise extreme caution when refilling the trap due to the caustic nature of the absorbent.
Dimensions:
- 100 cc Traps: 123/4 in. (32.4 cm) long x 13/4 in. (44 mm) diameter
- 250 cc Traps: 171/2 in. (44.4 cm) long x 2 in. (51 mm) diameter
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service