Skip to Content
Merck
All Photos(1)

Key Documents

4M9148-U

Supelco

Indoor Air Standard

50 components, certified reference material, 100 μg/mL each component in methanol: water (19:1)

Synonym(s):

Indoor Air Quality Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 3 × 1 mL

concentration

100 μg/mL each component in methanol: water (19:1)

technique(s)

gas chromatography (GC): suitable

format

multi-component solution

storage temp.

-10 to -25°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetone
    Benzene
    Bromodichloromethane
    Butyl acetate
    1-Butanol
    2-Butanone
    Chloroform
    Dibromochloromethane
    Decane
    Decanal
    1,4-Dichlorobenzene
    1,2-Dichloroethane
    Dichloromethane
    1,2-Dichloropropane
    2,4-Dimethylpentane
    Dodecane
    Ethylbenzene
    Ethyl alcohol, Pure
    Ethyl acetate
    2-Ethyltoluene
    3-Ethyltoluene
    4-Ethyltoluene
    Heptane
    Hexane
    Hexadecane
    (R)-(+)-Limonene
    Mesitylene
    4-Methyl-2-pentanone
    Nonanal
    Nonane
    Octane
    Pentadecane
    (1S)-(-)-alpha-Pinene
    (-)-beta-Pinene
    1-Propanol
    2-Propanol
    Styrene
    Tetrachloroethylene
    Tetradecane
    Durene
    Toluene
    Trichloroethylene
    Tridecane
    1,2,3-Trimethylbenzene
    1,2,4-Trimethylbenzene
    2,2,4-Trimethylpentane
    Undecane
    m-Xylene
    o-Xylene
    p-Xylene
    See All (50)

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service