Skip to Content
Merck
All Photos(1)

Key Documents

49018

Supelco

2,4′-DDT

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

storage temp.

2-30°C

Gene Information

mouse ... Esr1(13982)
rat ... Ar(24208)

General description

2,4′-DDT is an organochlorine pesticide, which can be widely used in agriculture for crop protection and disease vector control.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review on the usage of POP pesticides in China, with emphasis on DDT loadings in human milk
Wong.H.M, et al.
Chemosphere, 60(6), 740-752 (2005)
Assessment of the exposure to organochlorine pesticides, PCBs and PAHs in six species of predatory birds of the Canary Islands, Spain
Luzardo.PO, et al.
The Science of the Total Environment, 472, 146-153 (2014)
Vitellogenin?a biomarker for endocrine disruptors
Hansen.D.P, et al.
TrAC, Trends in Analytical Chemistry, 17(7), 448-451 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service