Skip to Content
Merck
All Photos(1)

Key Documents

47118

Supelco

Olive oil

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12164500

grade

analytical standard

Quality Level

description

refined

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.910 g/cm3

application(s)

food and beverages

format

neat

functional group

ester
oleic acid

shipped in

ambient

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

General description

Characterized olive oil samples are available for use as controls or check samples for fatty acid methyl ester (FAME) analyses. These reference materials can be used as qualitative standards. They provide an excellent means of standardizing the lipid procedures and comparing the results to others. Packed in an amber ampule under nitrogen.

Application

Olive oil may be used as an analytical reference standard for the determination of the analyte in milk samples by high performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

The health benefits of olive oil result from its higher proportion of monounsaturated fats, as well as its antioxidant constituents: hydroxytyrosol, oleuropein and oleocanthal.

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A (1998)
Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A (1998)
Analysis of human milk triacylglycerols by high-performance liquid chromatography with light-scattering detection.
Pons SM, et al.
Journal of Chromatography A (1998)
Nuno Rodrigues et al.
Food research international (Ottawa, Ont.), 128, 108759-108759 (2020-01-21)
Mediterranean olive heritage richness is poorly characterized. Olive oils from minor cultivars of Northeast-Portugal (Lentisca, Madural, Rebolã, Redondal, Verdeal and Verdeal Transmontana) from centenarian trees were chemical and sensory characterized, aiming to identify autochthonous cultivars capable of producing differentiated olive
Christos Bazakos et al.
Food chemistry, 134(4), 2411-2418 (2013-02-28)
Authenticity and traceability of high quality monovarietal extra virgin olive oils is a major concern for markets and consumers. Although analytical chemistry techniques are widely used to satisfy these needs recently developed DNA-based methods can serve as complementary approaches. A

Articles

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Learn more about Biodiesel as a renewable, alternative diesel fuel produced from vegetable oils, animal fats, or recycled restaurant grease.

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service