Skip to Content
Merck
All Photos(1)

Key Documents

47082-U

Supelco

EPA 8260 Internal Standards Mix HC

certified reference material, 10000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 8260
610

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

10000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 8260 Internal Standards Mix HC is a multi-component solution containing chlorobenzene-d5, 1,4-dichlorobenzene-d4, 1,4-difluorobenzene, and pentafluorobenzene as certified reference materials (CRMs).

US EPA 8000 Series methods are analytical methods for identifying and quantifying halogenated and aromatic volatile organic compounds (VOCs) on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Chlorobenzene-d5
    1,4-Dichlorobenzene-d4
    1,4-Difluorobenzene
    Pentafluorobenzene

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 2 - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service