Skip to Content
Merck
All Photos(1)

Key Documents

46937

Supelco

L-(+)-Lactic acid

analytical standard

Synonym(s):

(S)-2-Hydroxypropionic acid, Sarcolactic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O3
CAS Number:
Molecular Weight:
90.08
Beilstein:
1720251
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-negative bacteria

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

C[C@H](O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1

InChI key

JVTAAEKCZFNVCJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Salem Ibrahim et al.
International journal of molecular sciences, 20(14) (2019-07-19)
Cariogenic oral biofilms are strongly linked to dental caries around dental sealants. Quaternary ammonium monomers copolymerized with dental resin systems have been increasingly explored for modulation of biofilm growth. Here, we investigated the effect of dimethylaminohexadecyl methacrylate (DMAHDM) on the
Diamante Maresca et al.
Microorganisms, 7(7) (2019-07-18)
Phenotypic and genotypic evidence indicates that many LAB strains can grow in presence of oxygen and can shift from fermentative to aerobic and/or respiratory metabolism. The aerobic and respiratory growth of several LAB species have been studied, allowing the selection
Y Shirai et al.
Journal of medical entomology, 38(1), 51-54 (2001-03-28)
The attraction of Aedes albopictus (Skuse) to hands and forearms of human subjects treated with several concentrations of L-LA solution were studied in a test chamber containing proboscis-amputated mosquitoes. Fewer mosquitoes alighted on L-LA treated human skin than on water-treated
Mitsuharu Kaya et al.
Metabolism: clinical and experimental, 55(1), 103-107 (2005-12-06)
To investigate the effects of exercise on the plasma concentrations and urinary excretion of purine bases and oxypurinol, we performed 3 experiments with 6 healthy male subjects. The first was a combination of allopurinol intake (300 mg) and exercise (VO2max
J K Nicholson et al.
The Biochemical journal, 211(3), 605-615 (1983-06-01)
Spin echo Fourier transform proton n.m.r. spectra of whole blood contain resonances from both erythrocytes and plasma. A large number of well-resolved signals from mobile protons of low-molecular-weight metabolites in plasma and serum have been identified. Spectra from the plasmas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service