Skip to Content
Merck
All Photos(1)

Key Documents

46824-U

Supelco

EPA CLP Semivolatiles Surrogate Standards Mix

certified reference material, 4000 μg/mL each component in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

US EPA/CLP

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type surrogate

packaging

ampule of 1 mL

concentration

4000 μg/mL each component in methylene chloride

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA CLP Semivolatiles Surrogate Standards Mix is a multi-component solution containing certified reference materials (CRMs) such as 2-chlorophenol-3,4,5,6-d4, 1,2-dichlorobenzene-d4, 2-fluorobiphenyl, 2-fluorophenol, nitrobenzene-d5, phenol-d6, p-terphenyl-d14, and 2,4,6-tribromophenol.

US EPA Contract Laboratory Program (CLP) methods are analytical methods for identifying and quantifying organic compounds on the EPA′s Target Compound List (TCL) in groundwater, sediment, and soil samples at abandoned hazardous waste sites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    2-Chlorophenol-3,4,5,6-d4
    1,2-Dichlorobenzene-d4
    2-Fluorobiphenyl
    2-Fluorophenol
    Nitrobenzene-d5
    Phenol-d6
    p-Terphenyl-d14
    2,4,6-Tribromophenol
    See All (8)

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service