Skip to Content
Merck
All Photos(1)

Key Documents

442795

Supelco

tert-Butyl ethyl ether

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC(CH3)3
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731469
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor pressure

155 mmHg ( 25 °C)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.375 (lit.)

bp

72-73 °C (lit.)

mp

−97 °C (lit.)

density

0.742 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

CCOC(C)(C)C

InChI

1S/C6H14O/c1-5-7-6(2,3)4/h5H2,1-4H3

InChI key

NUMQCACRALPSHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyl ethyl ether is a fuel additive used in the formulation of automotive gasoline for reducing the emission of carbon monoxide and unburned hydrocarbons, and also to improve octane ratings.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation characteristic and products of ETBE (ethyl tert-butyl ether).
Liu X, et al.
Energy, 82, 184- 192 (2015)
Yuzo Asano et al.
Drug and chemical toxicology, 34(3), 311-317 (2011-06-09)
Ethyl tertiary-butyl ether (ETBE) is commonly used as an oxygenated gasoline additive. In this study, the prenatal developmental toxicity of ETBE was determined in rabbits. New Zealand white rabbits were given ETBE by gavage at 100, 300, or 1,000 mg/kg/day
Zuquan Weng et al.
Industrial health, 49(3), 396-399 (2011-03-05)
To clarify the genotoxicity of ethyl tertiary butyl ether (ETBE), a gasoline additive, male and female C57BL/6 mice of Aldh2+/+ and Aldh2-/- genotypes, aged 8 wk, were exposed to 0, 500, 1,750, or 5,000 ppm ETBE for 6 h/day, 5
Yoko Eitaki et al.
Journal of occupational health, 53(6), 423-431 (2011-10-15)
In order to measure occupational exposure concentrations of ethyl tertiary-butyl ether (ETBE), we developed a diffusive sampling method for monitoring ETBE and performed an ETBE exposure assessment. The applicability of diffusive samplers was examined by exposing the samplers to ETBE
Ann de Peyster
Birth defects research. Part B, Developmental and reproductive toxicology, 89(3), 239-263 (2010-06-15)
Ethyl t-butyl ether (ETBE) is a motor fuel oxygenate used in reformulated gasoline. Knowledge of developmental and reproductive toxicity potential of ETBE is critical for making informed decisions about acceptance and regulations. This review discusses toxicology studies providing information about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service