Skip to Content
Merck
All Photos(1)

Key Documents

442282

Supelco

2,3,4,6-Tetrachlorophenol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl4O
CAS Number:
Molecular Weight:
231.89
EC Number:
MDL number:
UNSPSC Code:
12000000
eCl@ss:
32150233
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Oc1c(Cl)cc(Cl)c(Cl)c1Cl

InChI

1S/C6H2Cl4O/c7-2-1-3(8)6(11)5(10)4(2)9/h1,11H

InChI key

VGVRPFIJEJYOFN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,3,4,6-Tetrachlorophenol belongs to the class of substituted phenol compounds, which are widely used in industrial applications such as synthesis of pesticides, dyes, drugs, plastics, etc. Due to their extensive usage, they are commonly found in environmental samples as pollutants.

Application

2,3,4,6-Tetrachlorophenol may be used as an analytical reference standard for the determination of the analyte in landfill leachates, aqueous samples, and environmental samples by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J S Karns et al.
Applied and environmental microbiology, 46(5), 1176-1181 (1983-11-01)
Resting cells of 2,4,5-trichlorophenoxyacetic acid-grown Pseudomonas cepacia AC1100 were able to completely and rapidly dechlorinate several chlorine-substituted phenols, including 2,4,5-trichlorophenol, 2,3,4,6-tetrachlorophenol, and pentachlorophenol. Several other trichlorophenols were only partially dechlorinated. The evidence suggests that 2,4,5-trichlorophenol is an intermediate in the
Acute and short-term toxicity of 2,3,4,6-tetrachlorophenol in rats.
M L Hattula et al.
Bulletin of environmental contamination and toxicology, 26(6), 795-800 (1981-06-01)
Pavel Smejtek et al.
The Journal of chemical physics, 120(3), 1383-1394 (2004-07-23)
We studied the effect of segmented solvent molecules on the free energy of transfer of small molecules from water into alkanes (hexane, heptane, octane, decane, dodecane, tetradecane, and hexadecane). For these alkanes we measured partition coefficients of benzene, 3-methylindole (3MI)
M K Männisto et al.
Water research, 35(10), 2496-2504 (2001-06-08)
The composition and chlorophenol-degrading potential of groundwater bacterial community in a permanently cold, oxygen-deficient chlorophenol contaminated aquifer at Kärkölä, Finland was studied with the aim of evaluating in situ bioremediation potential. The groundwater contained from 10(4) to 10(7) microscopically counted
J H Langwaldt et al.
Applied microbiology and biotechnology, 49(6), 663-668 (1998-07-31)
Air-lift percolator experiments simulated in situ subsurface degradation of 2,4,6-trichlorophenol, 2,3,4,6-tetrachlorophenol and pentachlorophenol, in mixtures and individually, by indigenous microorganisms from a chlorophenol-contaminated aquifer. Inoculation with a chlorophenol(CP)-degrading gram-positive isolate from the CP-contaminated groundwater did not significantly increase CP degradation

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service