44078-U
Octafluoronaphthalene
0.1 ng/mL in hexane, analytical standard
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
77101502
NACRES:
NA.24
Recommended Products
grade
analytical standard
packaging
pkg of 1 mL
concentration
0.1 ng/mL in hexane
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-30°C
General description
Octafluoronaphthalene is classified under the group of volatile organic compounds (VOCs).
Application
Octafluoronaphthalene may be used as an analytical reference standard for the quantification of the analyte in commercial samples using gas chromatography (GC) coupled to orthogonal time-of-flight (oa-TOF) mass spectrometry (MS).
It can be utilized as an internal standard for the analysis of polyfunctional thiols in wine samples by GC-MS.
It can be utilized as an internal standard for the analysis of polyfunctional thiols in wine samples by GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-14.8 °F - closed cup
Flash Point(C)
-26.0 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Compatibility of electron ionization and soft ionization methods in gas chromatography/orthogonal time-of-flight mass spectrometry.
Hejazi L, et al.
Rapid Communications in Mass Spectrometry, 23(14), 2181-2189 (2009)
Laura Mateo-Vivaracho et al.
Journal of chromatography. A, 1185(1), 9-18 (2008-02-09)
A fast method for the determination of aroma-powerful polyfunctional thiols at ng L(-1) level has been developed. Mercaptans are selectively retained in a 50mg solid-phase extraction cartridge and derivatization takes place in the same cartridge at room temperature (25 degrees
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service