Skip to Content
Merck
All Photos(1)

Key Documents

40439

Supelco

Methyl methacrylate solution

certified reference material, 1000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

O(C)C(=O)C(=C)C

InChI

1S/C5H8O2/c1-4(2)5(6)7-3/h1H2,2-3H3

InChI key

VVQNEPGJFQJSBK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-lasting allergic patch test reaction to methyl methacrylate.
Michael P Sheehan et al.
Dermatitis : contact, atopic, occupational, drug, 23(5), 242-243 (2012-09-27)
Bahram Mokhtari et al.
Journal of separation science, 36(2), 356-361 (2013-01-22)
This article describes the preconcentration of methyl methacrylate in produced water by the dispersive liquid-liquid microextraction using extraction solvents lighter than water followed by gas chromatography. In the present experiments, 0.4 mL dispersive solvent (ethanol) containing 15.0 μL extraction solvent
Garima Agrawal et al.
Clinical nuclear medicine, 38(1), 50-52 (2012-12-18)
Methyl methacrylate (MMA) pulmonary embolism is a serious potential complication of percutaneous vertebroplasty. We present a case of a 28-year-old man who presented to an outside institution with pleuritic chest pain after MMA vertebroplasty of the T11 thoracic vertebra for
Chunfeng Ma et al.
Colloids and surfaces. B, Biointerfaces, 100, 31-35 (2012-07-07)
Copolymerization of methyl methacrylate (MMA) and acrylate terminated poly(ethylene oxide-co-ethylene carbonate) (PEOC) macromonomer yields graft copolymer with PEOC side chains. By use of quartz crystal microbalance with dissipation (QCM-D), we have investigated the adsorption of proteins on the surface constructed
Nattika Saengkrit et al.
Carbohydrate polymers, 90(3), 1323-1329 (2012-09-04)
In this study, we examined the potential of cationic nanoparticle - polyethyleneimine-introduced chitosan shell/poly (methyl methacrylate) core nanoparticles (CS-PEI) for siRNA delivery. Initially, DNA delivery was performed to validate the capability of CS-PEI for gene delivery in the human cervical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service