Skip to Content
Merck
All Photos(1)

Key Documents

40077-U

Supelco

EPA 506 Phthalate Mix (7 component)

certified reference material, 1000 μg/mL each component in isooctane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 506

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-30°C

General description

EPA 506 Phthalate Mix (7 component) is a multi-component solution containing benzyl butyl phthalate, bis(2-ethylhexyl) adipate, bis(2-ethylhexyl) phthalate, dibutyl phthalate, diethyl phthalate, dimethyl phthalate, and di-n-octyl phthalate as certified reference materials (CRMs). The raw materials and solvents used in the preparation of EPA 506 Phthalate Mix (7 component) have been screened for identity and purity. The mixture is gravimetrically prepared and quantitatively analyzed by gas chromatography (GC).
US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment.

Application

EPA 506 Phthalate Mix (7 component) may be used as a CRM to quantify phthalate analytes in ambient and atmospheric air and emissions from polyvinyl chloride consumer goods by gas chromatography-mass spectrometry (GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Benzyl butyl phthalate
    Bis(2-ethylhexyl) adipate
    Bis(2-ethylhexyl) phthalate
    Dibutyl phthalate
    Diethyl phthalate
    Dimethyl phthalate
    Di-n-octyl phthalate

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The characteristics of atmospheric phthalates in Shanghai: a haze case study and human exposure assessment
Li Y, et al.
Atmospheric Environment, 178, 80-86 (2018)
A novel quantitation method for phthalates in air using a combined thermal desorption/gas chromatography/mass spectrometry application
Jo SH, et al.
Analytica Chimica Acta, 944, 29-36 (2016)
Characterization and flux assessment of airborne phthalates released from polyvinyl chloride consumer goods
Jo SH, et al.
Environmental Research, 165(1), 81-90 (2018)
Tandem malonate-based glucosides (TMGs) for membrane protein structural studies
Hussain H, et al.
Scientific reports, 7(1), 1-11 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service