Skip to Content
Merck
All Photos(1)

Key Documents

40069

Supelco

Dimethyl phthalate solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

InChI

1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3

InChI key

NIQCNGHVCWTJSM-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yiling Chen et al.
Journal of hazardous materials, 235-236, 92-100 (2012-08-14)
In this study, we demonstrate that the decomposition of dimethyl phthalate under microwave irradiation could be greatly enhanced over Fe@Fe(2)O(3) nanowires supported on activated carbon (Fe@Fe(2)O(3)/AC). The great enhanced decomposition of dimethyl phthalate could be attributed to a unique microwave
Wenxin Wang et al.
Environmental monitoring and assessment, 184(11), 6789-6798 (2011-12-08)
Atmospheric trace phthalic esters (PAEs) carried by total suspended particulates were systematically investigated. A total of 450 air samples were collected at six typical locations and three specific micro-environmental sites (a stadium with a new plastic track, new cars, and
L J Xu et al.
Ultrasonics sonochemistry, 20(3), 892-899 (2012-12-05)
A comprehensive study of the sonochemical degradation of dimethyl phthalate (DMP) was carried out using high-frequency ultrasonic processes. The effects of various operating parameters were investigated, including ultrasonic frequency, power density, initial DMP concentration, solution pH and the presence of
L J Xu et al.
Water research, 47(6), 1996-2004 (2013-02-12)
The merits of the combined process of high-frequency ultrasound (US) and catalyst-free ultraviolet irradiation (UV) have been evaluated in this study by investigating the sonophotolytic degradation of dimethyl phthalate (DMP). A 400 kHz ultrasonic system and a photolytic system at
Xing-hui Xia et al.
Huan jing ke xue= Huanjing kexue, 31(4), 969-975 (2010-06-10)
Irreversible sorption behavior of two phthalate acid esters (PAEs), Dimethyl phthalate (DMP) and Di-(2-ethyl-hexyl) phthalate (DEHP), on four natural sediment samples from the Yangtze River and the Yellow River has been studied by equilibrium sorption and multiple cycles of adsorption/desorption

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service