Skip to Content
Merck
All Photos(3)

Key Documents

29741-U

Supelco

Tenax®TA Thermal Desorption Tube

greener alternative

stainless steel, O.D. x I.D. x L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea

Synonym(s):

Tenax® TA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Tenax®TA, 60/80 mesh, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, preconditioned, pkg of 10 ea

material

stainless steel TD tube

Quality Level

Agency

EPA TO-1,IP-1B

description

sealed with (Brass Endcaps)

feature

preconditioned

packaging

pkg of 10 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax TA porous polymer

particle size

60/80 mesh

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Tenax® TA is preconditioned stainless steel that is compatible for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instrument. It is commonly used as a general adsorbent and efficiently adsorbs many non-polar compounds, such as hydrocarbons, pesticides, or biogenic volatile organic compounds (VOCs) due to its affinity for compounds with a wide range of boiling points from 60 to 300 °C.

It is also suitable for thermal desorption of volatiles because of its good thermal stability and hydrophobicity.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Application

Tenax® TA has been used as an absorbent for:
  • an active sampling of reduced sulfur compounds in ambient air
  • bioaccessibility experiments due to its infinite adsorption performance and recyclability

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of Uncertainty in Measurements of Reduced Sulfur Compounds by Active Sampling on Tenax TA{\textregistered} Tubes
Cecilia M, et al.
Chemical Engineering Journal, 54 (2016)
Comparison of two common adsorption materials for thermal desorption gas chromatography-mass spectrometry of biogenic volatile organic compounds.
Andrea M, et al.
Journal of Chromatography A, 1514, 16-28 (2017)
Evaluating the adsorption performance of Tenax TA{textregistered} in different containers: An isolation tool to study the bioaccessibility of nitro-PAHs in spiked soil.
Xianxian C, et al.
The Science of the Total Environment, 806, 150429-150429 (2022)

Related Content

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service