29104-U
Vials, open-top screw cap, 2 mL standard opening with closures, convenience pack, unassembled
clear glass vial, PTFE/silicone septum, pkg of × 100 ea
Synonym(s):
Open-top screw cap vials with closures
About This Item
Recommended Products
material
PTFE/silicone septum
clear glass vial
polypropylene cap
description
Unassembled
feature
closure type black polypropylene hole cap
closure type screw top vial
packaging
pkg of × 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mm
volume
2 mL
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
Related Categories
Suitability
Agilent
Agilent 1050
Hitachi
AS-1000, AS-2000, AS-4000
L7200/L7250
LEAP Technologies
CTC LC PAL, GC PAL
CombiPAL
HTC PAL/HTS PAL
PerkinElmer
Autosystem/AS-2000/XL
Clarus 500/600
Varian
CP-8410
8100/8200
9100/9090/9095
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service