2877605-U
Inlet Liner, Split/Splitless Type, Straight FocusLiner™ Design (wool packed)
pkg of 5 ea
Synonym(s):
GC accessory, glass wool, inlet liner
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115719
NACRES:
SB.54
Recommended Products
feature
Split/Splitless
packaging
pkg of 5 ea
L × O.D. × I.D.
95 mm × 5.0 mm × 3.4 mm
95 mm × 5.0 mm × 3.4 mm
compatibility
configured for Shimadzu 2010
Looking for similar products? Visit Product Comparison Guide
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
- Fits Shimadzu (GC-2010)
Application
- Split/Splitless Inlets in Gas Chromatography: What′s Up with All Those Different Glass Inlet Liners: This article reviews the various configurations of glass inlet liners used in split and splitless inlets for gas chromatography, focusing on their impact on sample introduction and analysis efficiency. The use of Straight FocusLiner Design (wool packed) inlets is highlighted for their ability to improve vaporization and reduce sample loss, making them essential in method development for accurate analytical results (Snow, 2022).
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service