27027-U
Vials, clear glass, screw top, graduated (vial only)
volume 22 mL, thread for 20-400, pkg of 100 ea
Synonym(s):
Graduated Clear Glass Vials
About This Item
Recommended Products
material
clear glass vial (with graduations)
description
cap not included
feature
closure type screw top vial
graduated
packaging
pkg of 100 ea
O.D. × H
23 mm × 85 mm
volume
22 mL
fitting
thread for 20-400
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service