27019
Assembled screw cap with hole with PTFE/silicone septum
black polypropylene hole cap, thread for 15-425, PTFE/silicone, septum thickness 1.5 mm, for use with 7 mL vial, pkg of 100 ea
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE/silicone
black polypropylene hole cap
feature
closure type black polypropylene hole cap
packaging
pkg of 100 ea
septum thickness
1.5 mm
fitting
thread for 15-425
compatibility
for use with 7 mL vial
Related Categories
Application
It may also be used in automated solid-phase microextraction-gas chromatography-mass selective detection (AutoSPME-GC-MSD) analyses for the determination of sugar-amino acid reaction mechanisms and production of branched-chain alkyl-substituted pyrazines.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
An Automated Solid-Phase Microextraction-Gas Chromatography-Mass Selective Detection Approach for the Determination of Sugar-Amino Acid Reaction Mechanisms.
Coleman WM and Lawson SN.
Journal of Chromatographic Science, 37 (10), 383-387 (1999)