Skip to Content
Merck
All Photos(1)

Key Documents

24815-U

Supelco

Merlin Microseal System (fits Agilent)

1 nut and 1 General Purpose (3-100 psi) septum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
eCl@ss:
32049213
NACRES:
SB.54
Pricing and availability is not currently available.

General description

The Merlin Microseal System is a septum-less system that provides very long life. Because there is not a septum to pierce, septum fragments will not be formed and deposited in the inlet liner. The septum contains a series of seals (wiper rib, o-rings, and a duckbill valve) that allow a needle to enter while maintaining a leak-free seal. The septum can be used only with a syringe that has a 23 gauge "blunt tipped" needle, or with an SPME fiber assembly with a 23 gauge needle. The thicker shaft is required to make the necessary contact with the septum seals.

Application

Merlin Microseal System may be used to replace traditional septum to avoid leaking and bleeding caused during operating GC/MS system.[1] It may also be equipped to the injection port during GC analysis for determination of plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater.[2]

Other Notes

Simply place the septum directly onto the septum cup and then add the nut (an additional adapter for the septum cup is not required for Agilent GCs). The septum incorporates a unique design with two sequential seals to provide a much longer life. Order replacement septa, or alternate versions, separately. Compatile with all Agilent autosamplers and stainless steel injection ports.

Note: Do not use with beveled tips.

Legal Information

MicroSeal is a trademark of Merlin Instrument Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of the plasticizer N-butylbenzenesulfonamide and the pharmaceutical Ibuprofen in wastewater using solid phase microextraction (SPME).
Huppert N
Fresenius Journal of Analytical Chemistry, 326 (6), 529-536 (1998)
J M Diserens
Journal of AOAC International, 84(3), 853-860 (2001-06-22)
Although the use of pentachlorophenol (PCP) is prohibited in most countries, it is still widely found in the wood of pallets, containers, crates and in cardboard, paper, etc. Such material may contain not only PCP but other chlorophenols as well.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service