23235
Amber storage bottles
volume 240 mL, O.D. × H 61 mm × 143 mm, thread for 24-400, bottle only, pkg of 12 ea
Synonym(s):
AMBER SCREW CAP BOTTLE ONLY 240ML PK12
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
amber glass bottle
feature
closure type screw top bottle
packaging
pkg of 12 ea
O.D. × H
61 mm × 143 mm
volume
240 mL
fitting
thread for 24-400
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage bottles are generally used to store stock solutions and other chemicals. Amber bottles are used to avoid oxidative changes in photosensitive chemicals.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Springer Science & Business Media, 06-Dec, 104-104 (2012)
Johanna Finn et al.
Cell reports, 26(11), 2942-2954 (2019-03-14)
Lung alveolar type I cells (AT1) and alveolar type II cells (AT2) regulate the structural integrity and function of alveoli. AT1, covering ∼95% of the surface area, are responsible for gas exchange, whereas AT2 serve multiple functions, including alveolar repair
Canola and Rapeseed: Production, Chemistry, Nutrition and Processing Technology
Shahidi, Fereidoon
Springer Science & Business Media, 06-Dec, 104-104 (2012)
Antonia E Schantl et al.
Nature communications, 11(1), 721-721 (2020-02-07)
Myo-inositol hexakisphosphate (IP6) is a natural product known to inhibit vascular calcification (VC), but with limited potency and low plasma exposure following bolus administration. Here we report the design of a series of inositol phosphate analogs as crystallization inhibitors, among
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service