22460
Supeltex M-4 Capillary Ferrule, General Purpose
I.D. 1.2 mm, 0.50-0.75 mm Column I.D., pkg of 10 ea
All Photos(1)
About This Item
UNSPSC Code:
41121815
packaging
pkg of 10 ea
I.D.
1.2 mm
compatibility
0.50-0.75 mm Column I.D.
General description
Supelco has refined the design of these graphite ferrules to provide a clean, sharp profile with minimal flash.
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Characteristics: Seals at one-quarter turn past fingertight. Maximum sealing surface contact, reduced risk of column contamination at installation.
Material Composition: flexible graphite
Maximum Temperature: 450 °C
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)