22004
Swagelok® Union
Swagelok® 400-6, brass, 1/4 in.
Synonym(s):
Swagelok® 400-6,BRASS UNION 1/4" PK2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115717
Recommended Products
material
brass
packaging
pkg of 2 ea
manufacturer/tradename
Swagelok® 400-6
fitting
1/4 in.
Looking for similar products? Visit Product Comparison Guide
General description
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic.
Nut and ferrules included.
Application
Brass Swagelok® union maybe used in determination of n-butylated trialkyllead compounds by gas chromatography with microwave plasma emission detection.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Marta Musioł et al.
Materials (Basel, Switzerland), 13(9) (2020-05-15)
The need for a cost reduction of the materials derived from (bio)degradable polymers forces research development into the formation of biocomposites with cheaper fillers. As additives can be made using the post-consumer wood, generated during wood products processing, re-use of
Determination of n-butylated trialkyllead compounds by gas chromatography with microwave plasma emission detection.
Scott E, at.al.
Analytical Chemistry, 54 (14), 2402-2405 (1982)
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service