Skip to Content
Merck

20411

Supelco

Glass Wool

Silanized, pkg of 50 g

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41121800

Pricing and availability is not currently available.

Agency

suitable for EPA 1621
suitable for EPA 1633
suitable for EPA OTM-45

packaging

pkg of 50 g

technique(s)

gas chromatography (GC): suitable

Looking for similar products? Visit Product Comparison Guide

General description

Glass wool is made up of short glass fibres and is non-flammable in nature. It is widely considered as a thermal insulation material in ships, automobiles, air conditioning ducts and water supply pipes. Furthermore, it is also used as noise absorbing material in concert halls, studios and movie theatres. Above all that it is non carcinogenic to humans.[1]
Wool plugs are often used in GC inlet liners to improve sample vaporization, and/or to keep non-volatile material from entering the column. They can also be used in packed GC columns, solvent desorption tubes, thermal desorption tubes, and purge traps to retain adsorbent beds.

Application

Silanized glass wool gas chromatographic packed column maybe used in an experimental procedure for measuring and collecting volatiles from edible oils.[2]

Other Notes

Silanized for general purpose use

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advanced Ceramic Technologies & Products. The Ceramic Society of Japan.
Imanaka Y.
Science, 18, 339-344 (1984)
Shayla Salzman et al.
Science advances, 6(24), eaay6169-eaay6169 (2020-06-26)
Most cycads engage in brood-site pollination mutualisms, yet the mechanism by which the Cycadales entice pollination services from diverse insect mutualists remains unknown. Here, we characterize a push-pull pollination mechanism between a New World cycad and its weevil pollinators that
Volatiles and oil quality.
Jackson HW and Giacherio DJ.
Journal of the American Oil Chemists' Society, 54 (10), 458-460 (1977)
Keunhong Jeong et al.
Magnetic resonance in chemistry : MRC, 56(11), 1089-1093 (2018-06-02)
Parahydrogen is a potentially significant source of hyperpolarization. However, a heat exchanger at an ultralow temperature, which is normally sustained wastefully using liquid nitrogen, is essential for the generation of hyperpolarized parahydrogen. In order to cut down on the use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service