Skip to Content
Merck
All Photos(1)

Key Documents

10343-U

Supelco

Amberlite IRN78 Ion Exchange Resin

hydroxide form, 16-50 mesh, bucket of 500 g

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52

product name

Amberlite IRN78 OH hydroxide form, bucket of 500 g

grade

nuclear grade

Quality Level

description

cross-linkage 8%

form

beads

packaging

bucket of 500 g

parameter

60 °C OH- form max. temp.

moisture

54-60%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

trimethylammonium

particle size

580-680 μm

operating pH

0-14

capacity

≥1.1 meq/mL by wetted bed volume

separation technique

anion exchange

General description

Amberlite IRN78, strong base anion exchange gel type polystyrene resin in hydroxide form, has quaternary ammonium functional group.
As a nuclear grade resin, a minimum of 95% of the available exchange sites are in the hydroxide form.

Application

Amberlite IRN78 hydroxide form was used as packing bed in vertical column chromatography.
Strongly basic (type I) anion exchange resin for water treatment, RADwaste, decontamination, etc.; useful for boron thermal regeneration.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of crude glycerol from transesterification rbd palm oil over homogeneous and heterogeneous catalyst for the biolubricant preparation
W. Isahak, M. Ismail, M. Yarmo, J. Jahim, J. Sali
Journal of Applied Sciences, 10, 2590-2595 (2010)
Non-destructive radioanalytical technique in characterization of anion exchangers Amberlite IRN78 and Indion H-IP
Singare, P. U.
J. Radioanal. Nucl. Chem., 299, 591-598 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service