Skip to Content
Merck
All Photos(1)

Key Documents

06846

Supelco

Chlorimuron ethyl

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Ethyl 2-(4-chloro-6-methoxy-2-pyrimidinylcarbamoylsulfamoyl)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H15ClN4O6S
CAS Number:
Molecular Weight:
414.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccccc1S(=O)(=O)NC(=O)Nc2nc(Cl)cc(OC)n2

InChI

1S/C15H15ClN4O6S/c1-3-26-13(21)9-6-4-5-7-10(9)27(23,24)20-15(22)19-14-17-11(16)8-12(18-14)25-2/h4-8H,3H2,1-2H3,(H2,17,18,19,20,22)

InChI key

NSWAMPCUPHPTTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Chlorimuron-ethyl is a member of sulfonylurea herbicides used wildly for pre- and post-emergence control of broad-leaved weeds in soybeans, maize, peanuts, beans, and other crops. It is characterized by very low application rates, high herbicidal activity, good crop selectivity, and low toxicity.

Chlorimuron-ethyl is an acetolactate synthase (ALS) inhibitor, that acts by inhibiting the biosynthesis of the essential amino acids like valine, leucine, and isoleucine. This stops the plant growth as cell division of growing roots and shoots tips are inhibited.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Chlorimuron-ethyl CRM may also find following uses:
  • To develop and validate an RP-HPLC method for the determination of metsulfuron-methyl, chlorantraniliprole, and chlorimuron-ethyl residues in stems of Oryza sativa
  • To study the rate of biodegradation of chlorimuron-ethyl using Pseudomonas sp. LW3 in liquid culture and contaminated soil
  • To investigate the toxicological effects of chlorimuron-ethyl, cadmium, and copper on germination rate and shoot and root elongation of wheat Triticum aestivum
  • Study the effect of chlorimuron-ethyl on physiological mechanisms in wheat and identify valuable biochemical biomarkers for the pollution of chlorimuron-ethyl in soil-plant systems
  • Study the biochemical responses to joint stress of chlorimuron-ethyl and cadmium in wheat Triticum aestivum
  • To study the photolysis of chlorimuron-ethyl on a soil surface, under sunlight and UV light
  • To determine the photodegradation kinetics of chlorimuron-ethyl in an aqueous solution under sunlight and UV light

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hailian Zang et al.
Chemosphere, 144, 176-184 (2015-09-13)
In this study, the effects of cultivation conditions on the degradation of chlorimuron-ethyl by Stenotrophomonas maltophilia D310-3, which exhibits a high chlorimuron-ethyl-degrading capability, were investigated. To improve the biodegradation efficiency, the cultivation conditions were optimized using response surface methodology (RSM)
Mei-E Wang et al.
Environmental pollution (Barking, Essex : 1987), 144(2), 572-580 (2006-03-15)
Biochemical responses to joint stress of chlorimuron-ethyl and cadmium (Cd) in wheat Triticum aestivum were examined. The joint action of chlorimuron-ethyl and Cd weakened the inhibition of Cd or chlorimuron-ethyl on the formation of chlorophyll. It was deduced that wheat
Meie Wang et al.
Ecotoxicology and environmental safety, 64(2), 190-197 (2005-06-01)
Changes in the activity of antioxidant enzymes including peroxidases (POD) and superoxide dismutases (SOD) and in the contents of chlorophyll (CHL), malondialdehyde (MDA), and soluble proteins (SP) in wheat (Triticum aestivum) under the stress of chlorimuron-ethyl in soil were examined.
Meie Wang et al.
Ecotoxicology and environmental safety, 60(2), 169-175 (2004-11-18)
Investigation of the toxicological effects of some agricultural pollutants on germination rate and on shoot and root elongation of wheat (Triticum aestivum) was carried out. Seeds of wheat were exposed to various concentrations of chlorimuron-ethyl with or without cadmium and
Ji-Ping Ma et al.
FEMS microbiology letters, 296(2), 203-209 (2009-05-23)
The chlorimuron-ethyl-degrading bacterium LW3 was isolated from contaminated soil and identified by 16S rRNA gene sequencing as Pseudomonas sp. When chlorimuron-ethyl was provided as the sole nitrogen source, the degradation efficiency in liquid medium was about 81.0% after 7 days

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service