Z764892
CryoFile® freezer box, for batching and storing cryogenic vials or ampules
white
Synonym(s):
freezer containers, storage boxes, storage containers
About This Item
material
cardboard (Water repellent)
feature
Holds 81 x 1.2-2 mL tubes
packaging
pkg of 15 ea
manufacturer/tradename
WHEATON® W651603
technique(s)
cryopreservation: suitable
L × W × H
130 mm × 130 mm × 52 mm
color
white
Looking for similar products? Visit Product Comparison Guide
General description
The box withstands cryogenic temperatures to 196°C.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service