Z764108
MicroTissues® 3D Petri Dish® micro-mold mixed spheroid kit
3 small and 3 large, fits 24 well plates
Synonym(s):
3D, 3D Cell Culture
About This Item
Recommended Products
material
spherical
sterility
sterile; autoclaved
feature
lid: no
manufacturer/tradename
MicroTissues Inc. 24-1
packaging
pack of (Contains 3 small and 3 large)
1 of 4
This Item | Z764094 | Z764116 | Z764043 |
---|---|---|---|
spherical | spherical | spherical | spherical |
MicroTissues Inc. 24-1 | MicroTissues Inc. 12-1 | MicroTissues Inc. 8-1 | MicroTissues Inc. 24-96 |
lid: no | lid: no | lid: no | lid: no, 8 x 12 array |
pack of (Contains 3 small and 3 large) | pack of (Contains 3 small and 3 large) | pack of 8 ea | pack of 6 ea |
General description
Since the agarose is transparent, the spheroids or microtissues that form at the bottom of each agarose micro-well can be easily viewed using a standard inverted microscope using phase contrast, bright field or fluorescence microscopy.
Features and Benefits
- The micro-molds are reusable up to 12 times and can be sterilized via a standard steam autoclave (30 min, dry cycle).
- The micro-molds should be stored in a covered container to maintain sterility and to avoid collecting dust or fibres on their small features.
- Any microscope with sufficient magnification and resolution functions for cell based views or images will be suitable for use with the 3D Petri Dish products.