Skip to Content
Merck
All Photos(1)

Key Documents

Z744194

eXTReme SealMate System for Adhesive Microplate-sealing Films

new

eXTReme starter kit, includes SealMate dispenser + 2 rolls of eXTReme PCR film, non-sterile

Synonym(s):

microplate sealing film, multiwell plate sealing film, sealing film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410

sterility

non-sterile

feature

DNase free
RNase free

packaging

pkg of 1 kit (Dispenser plus 2 rolls of 80 sheets per roll)

manufacturer/tradename

Excel Scientific SM-KIT-XTR

parameter

-20-+120 °C temp. range

technique(s)

PCR: suitable

General description

Extra strong adhesive minimizes evaporation but removes without residue. The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films. Ideal for use under the hood; the SealMate dispenser minimizes film contamination. Each sheet is separated by an adhesive-free, perforated, color-coded blue band. The smooth lower surface of the dispenser can be used to seal the film to the plate. The lower support prop prevents dispensing bill from contacting surfaces and protects unused seals from contamination. Replacement rolls can be easily installed by releasing the locking clasps on the dispenser

Application

PCR, General Storage

Features and Benefits

  • Extra strong adhesive minimizes evaporation yet removes easily without leaving residue on the plate.
  • Each sheet is separated by an adhesive-free, perforated, color-coded blue band for easy seperation.
  • The smooth surface of the dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films

Legal Information

SealMate is a trademark of Excel Scientific, Inc.
eXTReme is a trademark of Excel Scientific, Inc.

replacement part


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service