Z744089
Autoclave indicator tape
W × L 3/4 in. × 2,160 in.
Synonym(s):
Indicator tape
About This Item
Recommended Products
packaging
pack of 1 roll
manufacturer/tradename
Diversified Biotech ACTP-1000
W × L
3/4 in. × 2,160 in.
Looking for similar products? Visit Product Comparison Guide
1 of 4
This Item | SAB2102412 | SAB5700877 | SAB4502952 |
---|---|---|---|
rabbit | rabbit | rabbit | rabbit |
buffered aqueous solution | buffered aqueous solution | liquid | buffered aqueous solution |
immunoblotting: 4-8 μg/mL using human HeLa cells extract, immunofluorescence: 2.5-5 μg/mL using mouse embryo fibroblast NIH-3T3 cells | western blot: suitable | immunohistochemistry: 1:50-1:200, western blot: 1:500-1:2000 | ELISA: 1:20000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000 |
−20°C | −20°C | −20°C | −20°C |
dry ice | wet ice | wet ice | wet ice |
General description
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Need A Sample COA?
This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service