Key Documents
Z743330
ContainerSEAL® XL sealing tape
W × L 1 in. × 108 ft, white
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech CSEAL-XLW
W × L
1 in. × 108 ft
color
white
General description
Enjoy all the benefits of PetriSEAL and ContainerSEAL in a new
wider format. ContainerSEAL XL makes it easier to seal large
containers and bottles and to cover wider gaps ensuring a tight
seal.
Legal Information
ContainerSEAL is a registered trademark of Diversified Biotech