Skip to Content
Merck
All Photos(2)

Key Documents

Z740303

Disposable Plastic BioSafe Pipette

capacity 25 mL

Synonym(s):

barrier filter, barrier filter pipette, pipet, pipette, plugged pipette, serological, serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

Pricing and availability is not currently available.

material

plastic (Disposable)

sterility

sterile

feature

graduations Subdivisions 0.1mL
plugged

packaging

pack of 150 ea

manufacturer/tradename

Cole Parmer 1300254

graduations and accuracy measuring range

0.1 mL

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
Z740301Z740302Z740307
feature

graduations Subdivisions 0.1mL, plugged

feature

graduations Subdivisions 0.1mL, plugged

feature

graduations Subdivisions 0.1mL, plugged

feature

graduations Subdivisions 0.5mL, plugged

material

plastic (Disposable)

material

plastic (Disposable)

material

plastic (Disposable)

material

plastic (Disposable)

manufacturer/tradename

Cole Parmer 1300254

manufacturer/tradename

Cole Parmer 1300252

manufacturer/tradename

Cole Parmer 1300253

manufacturer/tradename

Cole Parmer 2299932

capacity

25 mL

capacity

5 mL

capacity

10 mL

capacity

25 mL

packaging

pack of 150 ea

packaging

pack of 200 ea

packaging

pack of 200 ea

packaging

pack of 100 ea

General description

Eliminate Cross Contamination
The first pipette with total contamination control


The Argos BioSafe pipette is a standard serological pipette with a unique barrier filter. If over-aspiration occurs with BioSafe pipettes, liquid is “locked” and unable to pass through to filter. Similar in theory to a barrier pipette tip, BioSafe pipettes provide sample integrity while promoting user safety. It also protects electronic pipettors from contamination and cross contaminating samples by over aspiration.

Ideal in applications involving any potentially hazardous or dangerous biological fluids, infectious diseases or when cross over contamination is an issue.

  • Unique filter plug “locks” liquid in pipette during over-aspiration
  • Available in 5, 10 & 25 mL (individually wrapped)
  • Eliminate aerosol or liquid contamination
  • Total contamination protection for pipettor and samples
  • Use in place of filtered pipette tips

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service