Z726710
Spectra-Por® Float-A-Lyzer® G2
red, 1 mL, MWCO 20 kDa
Synonym(s):
Float-A-Lyzer G2, G2 Float-A-Lyzer, Spectra-Por G2
All Photos(3)
About This Item
UNSPSC Code:
41123100
NACRES:
NB.22
Recommended Products
material
red
packaging
pack of 12 ea (dry with glycerol)
manufacturer/tradename
Spectrum Labs G235033
pore size
20 kDa MWCO
capacity
1 mL
Related Categories
General description
- 95 - 98% Sample Recovery
- Highest Membrane Purity
- Superior Handling & Leak Prevention
- Volume Specific Dilution Control
- Complete & Ready-to-Use Packaging
Spectrum′s proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 4 precise MWCO′s (color coded for MWCO variants) with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval. Only the Float-A-Lyzer® G2 assures a 95-98% sample recovery while maintaining 99% sample purity and <1% sample dilution.
The leak-proof screw-on cap with sealing o-ring provides easy access with included pipette for loading, in-process testing and sample retrieval, without the risk of needle punctures. The included floatation ring improves sample buoyancy and vertical orientation during dialysis. The sleek design allows multiple samples to be dialyzed in the same reservoir.
No additional kits, buoys or accessories are required. The Float-A-Lyzer G2 is packaged dry and ready-to-go with all components included. Simply rinse with water, load the sample with a pipette, and dialyze.
Spectrum Laboratories, Inc. introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation of Ready-to-Use laboratory dialysis devices featuring a Ultra-pure Biotech Cellulose Ester (CE) Membrane
Legal Information
Float-A-Lyzer is a registered trademark of Spectrum Laboratories, Inc.
Spectra/Por is a registered trademark of Spectrum Laboratories, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)