Z711063
Greiner 96 well plates, polypropylene
size 96 well, skirt
Synonym(s):
96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates
About This Item
material
colorless wells
polypropylene
sterility
non-sterile
feature
skirt
packaging
case of 40
manufacturer/tradename
Greiner 652270
size
96 well
working volume
200 μm
color
clear
clear colorless plate
application(s)
agriculture
agriculture
binding type
non-treated surface
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Suitability
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service