Key Documents
Z707643
TPP® cell culture spatula
Synonym(s):
cell scrapers, cell spatula
About This Item
UNSPSC Code:
41122103
Pricing and availability is not currently available.
material
polypropylene
sterility
sterile
packaging
pack of 1
case of 150
manufacturer/tradename
TPP 99010
blade L × W
195 mm × 14 mm , fixed blade
size
suitability
suitable for (tissue culture dish 150, various other cell culture applications)
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
CLS3010CLS3011Z707805
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
packaging
pack of 1
packaging
case of 100
packaging
case of 100
packaging
case of 72
manufacturer/tradename
TPP 99010
manufacturer/tradename
Corning 3010
manufacturer/tradename
Corning 3011
manufacturer/tradename
TPP 92424
blade L × W
195 mm × 14 mm , fixed blade
blade L × W
-
blade L × W
-
blade L × W
-
size
size
-
size
-
size
24 wells
General description
The TPP cell spatula is mainly used with TPP tissue culture flasks with re-closable lid or peel off foil and with the large TPP tissue culture dish.
The advantages are:
  • Special formed blade with sharp edges
  • Small raised knobs on the handle
  • Lint-free blister packaging
  • User friendly TPP dispenser case
Legal Information
TPP is a registered trademark of TPP Techno Plastic Products AG