Key Documents
Z614017
Vivaspin 500 centrifugal concentrators
MWCO 5 kDa
About This Item
UNSPSC Code:
41104916
NACRES:
NB.24
Pricing and availability is not currently available.
material
polycarbonate housing
polyethersulfone membrane
description
PES membrane
sterility
non-sterile
feature
holdup volume 5 μL
packaging
package of 100 ea
manufacturer/tradename
Sartorius VS0112
parameter
15000 × g max. centrifugal force
dead stop volume
5 μL
membrane area
0.5 cm2
tube size
~2.2 mL
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
Z614084Z614033Z614068
material
polycarbonate housing, polyethersulfone membrane
material
polycarbonate housing, polyethersulfone membrane
material
polycarbonate housing, polyethersulfone membrane
material
polycarbonate housing, polyethersulfone membrane
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
Sartorius VS0112
manufacturer/tradename
Sartorius VS0132
manufacturer/tradename
Sartorius VS0102
manufacturer/tradename
Sartorius VS0122
working volume
500 μL
working volume
500 μL
working volume
500 μL
working volume
500 μL
packaging
package of 100 ea
packaging
package of 100 ea
packaging
package of 100 ea
packaging
package of 100 ea
General description
  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.