Z601160
Hettich® MIKRO 220/220R centrifuge accessories
for MIKRO 220R Centrifuge, Adapter for 15 mL Falcon tubes (17x120 mm)
All Photos(1)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
Recommended Products
feature
Holds 15 mL tubes
packaging
pack of 6 ea
manufacturer/tradename
Hettich 1631
technique(s)
centrifugation: suitable
compatibility
for use with 1016, 1137 (rotors)
Related Categories
General description
- compatible with rotor 1016 (Z601381)
- rotor 1016 (Z601381) is only compatible with the MIKRO 220R Centrifuge
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
required but not provided
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)