Z370843
Scienceware® pipette reservoir, 12 well
each well holds 5 mL and has a V-bottom
Synonym(s):
multichannel pipette reservoir, reagent reservoir
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121522
NACRES:
NB.01
Recommended Products
material
V-bottom bottom
polypropylene
white
sterility
sterile
packaging
pack of 8 ea
manufacturer/tradename
Bel-Art F37880-0000
capacity
60 mL
General description
For use with multi-channel pipettes, to deliver a different solution to each row of a multiwell plate. Minimizing waste, this 12 Channel Reagent Reservoir′s V-shaped wells allow complete sample withdrawal.
- Each channel holds 5ml; numbered to facilitate serial dilution
- Chemical resistant polypropylene construction; fitted lid included
- Can be used with single, 8 or 12 channel pipettors
- Wide profile provides added stability; measures 12.7cm L x 5.8cm W (5” x 2¼”)
- Individually sealed and sterilized; autoclavable at 121ºC (250ºF)
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service