Z364495
Nalgene® media bottles, style 2019
capacity 125 mL
Synonym(s):
bottle nalgene, nalgene containers, sterile bottles
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
high-density polyethylene cap
square bottle
transparent PETG
Joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations
with cap
packaging
pack of 48 ea (2 x pack of 24)
manufacturer/tradename
Nalgene® 2019-0125
capacity
125 mL
Related Categories
General description
- Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
- Designed for storing buffers, cultures, and culture media for various laboratory applications.
- Features a leakproof closure for sample protection.
- Heat-shrink band around closure and neck facilitate tamper-evident seal.
- Ideal for refrigerated/frozen storage down to -40°C.
Media bottles (square), with white HDPE screw closure. Low gas permeability, non-pyrogenic. Heat-shrink band around closure and neck provides tamper-evident seal.
Features and Benefits
- Avoids depyrogenation, expensive washing, and autoclaving steps
- Sterile and non-pyrogenic
- Obtained in low particulate format
- Validated by extensive documentation
- Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
- Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Already Own This Product?