Key Documents
Z336793
Workstation storage/dispenser box, PMMA
small
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
Pricing and availability is not currently available.
manufacturer/tradename
BRAND
L × W × H
165 mm × 152 mm × 178 mm
Compare Similar Items
View Full Comparison
Show Differences
1 of 4
This Item
Z336785HS234634HS120668
L × W × H
165 mm × 152 mm × 178 mm
L × W × H
165 mm × 152 mm × 355 mm
L × W × H
154 mm × 171 mm × 206 mm
L × W × H
123 mm × 172 mm × 99 mm
General description
Ideal for storage and dispensing many small items used in the lab like disposable pipette tips, microcentrifuge tubes, pasteur pipette etc. The top cover is removable and the front cover is hinged.
Other Notes
BR131902 (India)