Skip to Content
Merck
All Photos(1)

Key Documents

Y1250

Sigma-Aldrich

Yeast Nitrogen Base With Amino Acids

Non-selective yeast growth medium

Synonym(s):

ynb with amino acids

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.

grade

for molecular biology

Quality Level

sterility

non-sterile

form

powder

technique(s)

microbiological culture: suitable

pH

5.3-5.5(0.67% solution)

application(s)

microbiology

storage temp.

room temp

suitability

nonselective for Candida spp.
nonselective for Pichia spp.
nonselective for Saccharomyces spp.
nonselective for Zygosaccharomyces spp.

General description

Yeast Nitrogen Base is a highly-referenced growth medium used for the cultivation of yeast. This nutrient-rich microbial broth contains amino acids, nitrogen, vitamins, trace elements and salts.

Application

Yeast Nitrogen Base With Amino Acids has been used:
  • in the ZYP-31 media preparation for E.coli[1]
  • in synthetic dextrose (SD) medium to evaluate the influence of nitrogen source[2]
  • to culture Candida utilis for the production of 13C-labeled yeast extract as internal standard[3]
  • in the preparation of imaging medium[4]

Suitable for use in classifying yeasts based on carbon requirements. Also suitable for general growth of yeasts with the addition of a carbon source (usually glucose).Yeast nitrogen base with amino acids is a satisfactory nitrogen source for general growth media. Contains all essential nutrients required for the growth of yeasts except carbohydrate.

Components

Component, amount (μg/L unless indicated)

Nitrogen Sources:
Ammonium sulfate, 5.0 g/L

Amino Acids:
L-histidine, 10 mg/L
DL-methionine, 20 mg/L
DL-tryptophan, 20 mg/L

Vitamins:
Biotin, 2.0
Calcium pantothenate, 400
Folic acid, 2.0
Inositol, 2.0 mg/L
Nicotinic acid, 400
p-Aminobenzoic acid, 200
Pyridoxine HCl, 400
Riboflavin, 200
Thiamine HCL, 400

Trace Elements:
Boric acid
Copper sulfate
Potassium iodide
Ferric chloride
Magnesium sulfate
Sodium molybdate
Zinc sulfate

Salts:
Potassium phosphate monobasic, 1.0 g/L
Magnesium sulfate, 0.5 g/L
Sodium chloride, 0.1 g/L
Calcium chloride, 0.1 g/L

Reconstitution

Make a 10x concentrated stock solution by stirring to suspend 6.7g yeast nitrogen base powder and 5 g glucose in 100 mL water. Warm if necessary to aid solubilization. Filter sterilize and store at 2-8°C.

Dilute the 10x concentrated stock to a 1x working solution by adding 100mL concentrated stock to 900mL sterile water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Live bio-imaging with fully bio-compatible organic fluorophores
Yang I, et al.
Journal of Photochemistry and Photobiology. B, Biology, 166, 52-57 (2017)
Renée Hermanns et al.
Journal of fungi (Basel, Switzerland), 5(3) (2019-09-12)
Recurrent vulvovaginal candidiasis (RVVC) is predicted to increase to almost 158 million cases annually by 2030. Extensive self-diagnosis and easily accessible over-the-counter (OTC) fungistatic drugs contribute to antifungal-resistance, illustrating the need for novel therapies. Honey possesses multiple antimicrobial mechanisms, and
New perspectives in shake flask pH control using a 3D-printed control unit based on pH online measurement
Ude C, et al.
Sensors and Actuators B, Chemical, 221, 1035-1043 (2015)
Corinne Maufrais et al.
G3 (Bethesda, Md.) (2021-04-07)
The genomes of a large number of Cryptococcus neoformans isolates have been sequenced and analyzed in recent years. These genomes have been used to understand the global population structure of this opportunistic pathogen. However, only a small number of South
Fast filtration of bacterial or mammalian suspension cell cultures for optimal metabolomics results
Bordag N, et al.
PLoS ONE, 11(7), e0159389-e0159389 (2016)

Protocols

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Yeast culture techniques: Model systems for eukaryotic studies with liquid media or agar plate growth.

Questions

1–3 of 3 Questions  
  1. Is the Yeast nitrogen base with amino acids food grade or FCC

    1 answer
    1. This item is intended for research use only and is not suitable as food grade or FCC material. Currently, there is no product option available that meets this requirement.

      Helpful?

  2. Is there a technical data sheet avaliable for product Y1250? I would like to know the concentrations of trace elements.

    1 answer
    1. The concentration of the trace elements is not reported on the datasheet. The trace elements in the formulation are as follows:

      - Boric Acid: 500.0µg
      - Potassium Iodide: 100.0µg
      - Manganese Sulfate: 400.0µg
      - Zinc Sulfate: 0.4mg
      - Calcium Chloride: 0.1g
      - Copper Sulfate: 40.0µg
      - Ferric Chloride: 200.0µg

      Helpful?

  3. Hi, Is this product Kosher and Halal certified? And food grade? Y1250 - Sigma-Aldrich Yeast Nitrogen Base With Amino Acids Kr, Benedicte

    1 answer
    1. This product is intended for research use only and is not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses, or any type of consumption or application to humans or animals. Currently, a food-grade product is not offered.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service