Skip to Content
Merck
All Photos(3)

Key Documents

XNAP2

Sigma-Aldrich

Extract-N-Amp Plant PCR Kit

sufficient for 100 extractions, sufficient for 100 amplifications

Synonym(s):

Plant direct PCR kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106303
NACRES:
NA.55

usage

sufficient for 100 amplifications
sufficient for 100 extractions
sufficient for 100 reactions

feature

dNTPs included
hotstart

technique(s)

PCR: suitable

color

colorless

application(s)

agriculture

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

The Extract-N-Amp Plant PCR Kits are intended for direct PCR (polymerase chain reaction). This kit contains all the reagents necessary to rapidly extract genomic DNA from plant leaves and amplify targets of interest by PCR. It is a novel Extraction Solution that eliminates the need for conventional freezing of plant tissues.

Application

Extract-N-Amp Plant PCR Kit has been used for genomic DNA isolation from plants and the fungal mycelium. It has also been used in polymerase chain reaction (PCR).
Extract-N-Amp Plant PCR Kit is suitable for use in genotyping.

It has been used forDNA extraction from fresh mushrooms.

Features and Benefits

  • Plant genomic DNA can be extracted for PCR in under 15 minutes using a method.
  • No freezing, mechanical disruption, organic extraction, column purification or precipitation is necessary
  • A specially formulated PCR ReadyMix is provided for use with the extracted DNA
  • The Hot Start antibody ensures highly specific PCR amplification of genomic DNA
  • Suitable for high-throughput plant genetic analysis.
  • Lengthy enzymatic digestions are not required, saving time and effort.
  • The extracted DNA remains stable at 4 °C for atleast 6 months.

Components

Extract-N-Amp Plant PCR Kits contain Dilution solution, Extraction solution, and Extract-N-Amp PCR Reaction Mix (2X) (buffer, salts, dNTPs, Taq polymerase, and JumpStart Taq antibody)

Other Notes

For additional information, please see www.sigma-aldrich.com/extract-n-amp.
The Extract-N-Amp Plant PCR Kits are for laboratory use only. Not for drug, household, or other uses.

Legal Information

Extract-N-Amp is a trademark of Sigma-Aldrich Co. LLC
ReadyMix is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ante Karoglan et al.
Acta dermato-venereologica, 99(13), 1253-1257 (2019-10-02)
Imbalance in skin microflora, particularly related to certain Cutibacterium acnes strains, may trigger acne. Application of non-acne-causing strains to the skin may modulate the skin microbiome and thereby lead to a reduction in acne. This pilot study evaluates the safety
Elin Videvall et al.
mSystems, 2(6) (2017-11-29)
The gut microbiome of animals is emerging as an important factor influencing ecological and evolutionary processes. A major bottleneck in obtaining microbiome data from large numbers of samples is the time-consuming laboratory procedures required, specifically the isolation of DNA and
Phylogenetic placements and cultural characteristics of Tuber species isolated from ectomycorrhizas
Obase K, et al.
Mycoscience (2021)
Matthew E Smith et al.
PloS one, 8(1), e55160-e55160 (2013-02-06)
Ectomycorrhizal (ECM) plants and fungi can be diverse and abundant in certain tropical ecosystems. For example, the primarily paleotropical ECM plant family Dipterocarpaceae is one of the most speciose and ecologically important tree families in Southeast Asia. Pakaraimaea dipterocarpacea is
Len N Gillman et al.
Journal of evolutionary biology, 23(6), 1327-1330 (2010-05-12)
A faster rate of nuclear DNA evolution has recently been found for plants occupying warmer low latitudes relative to those in cooler high latitudes. That earlier study by our research group compared substitution rates within the variable internal transcribed spacer

Articles

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Extract-N-Amp kits extract and amplify genomic DNA, optimized for plant tissue without inhibition concerns.

Protocols

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

The Extract-N-Amp™ Plant PCR Kits contain all of the reagents required to rapidly extract and amplify genomic DNA from plant leaves.

Related Content

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Explore technology and reagent portfolios for plant breeding workflows, accelerating the development of new crop varieties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service