Skip to Content
Merck
All Photos(3)

Key Documents

WTA1

Sigma-Aldrich

TransPlex® Whole Transcriptome Amplification Kit

DNA polymerase separate.

Synonym(s):

Transcriptome Amplification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NA.55

Quality Level

technique(s)

whole genome amplification: suitable

shipped in

wet ice

storage temp.

−20°C

General description

TransPlex®, a Whole Transcriptome Amplification (WTA) method, allows for representative amplification of nanogram quantities of total RNA in less than 4 hours without 3′-bias. Microgram quantities of amplification product generated from tissue, cultured cells, formalin-fixed samples, or serum are suitable for downstream applications such as qPCR and microarray analyses.

Application

TransPlex® Whole Transcriptome Amplification Kit has been used to synthesize double-stranded cDNA. It has also been used in the amplification of cDNA.
Suitable for use with downstream applications including:
  • qPCR
  • microarray analysis
  • cloning

Features and Benefits

  • Amplification of total RNA in less than 4 hours with less than 30 minutes of "hands-on" time
  • Only 5 ng of starting material required to produce a highly representative library from total RNA
  • Microgram quantities of amplification product generated from intact RNA from tissue, cultured cells, serum, or degraded RNA from formalin-fixed paraffin-embedded samples

Principle

The WTA process involves two steps. In the first step, sample RNA is reverse transcribed with non-self-complementary primers composed of a quasi-random 3′ end and a universal 5′ end. As polymerization proceeds, displaced single strands serve as new templates for primer annealing and extension. The resultant OmniPlex cDNA library, comprised of random, overlapping 100 - 1000 base fragments flanked by universal end sequence, is then amplified by PCR with the universal primer to produce WTA product.

Legal Information

TransPlex is a registered trademark of Rubicon Genomics, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • W4502Water, Nuclease-Free Water, for Molecular BiologySDS

  • D7295Deoxynucleotide Mix, 10 mM, Molecular Biology ReagentSDS

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Munehiro Okamoto et al.
Scientific reports, 5, 8850-8850 (2015-03-07)
We discovered a lethal hemorrhagic syndrome arising from severe thrombocytopenia in Japanese macaques kept at the Primate Research Institute, Kyoto University. Extensive investigation identified that simian retrovirus type 4 (SRV-4) was the causative agent of the disease. SRV-4 had previously
Catarina Guimarães-Teixeira et al.
Journal of personalized medicine, 11(10) (2021-10-24)
(1) Background: Methylation of N6-adenosine (m6A) is the most abundant messenger RNA (mRNA) modification in eukaryotes. We assessed the expression profiles of m6A regulatory proteins in renal cell carcinoma (RCC) and their clinical relevance, namely, as potential biomarkers. (2) Methods:
Sa Xiao et al.
Virus research, 145(1), 80-91 (2009-06-23)
The complete genome sequence of avian paramyxovirus serotype 7 (APMV-7) prototype strain dove/Tennessee/4/75 was determined. The genome size is 15,480 nucleotides (nt) long and follows the "rule of six". The genome contains six non-overlapping genes in the order of 3'-N-P/V/W-M-F-HN-L-5'.
Anne H Rowley et al.
The Journal of infectious diseases, 203(7), 1021-1030 (2011-03-16)
Intracytoplasmic inclusion bodies (ICI) have been identified in ciliated bronchial epithelium of Kawasaki disease (KD) patients using a synthetic antibody derived from acute KD arterial IgA plasma cells; ICI may derive from the KD etiologic agent. Acute KD bronchial epithelium
Ki Wook Kim et al.
Open forum infectious diseases, 6(2), ofz025-ofz025 (2019-03-01)
The importance of gut bacteria in human physiology, immune regulation, and disease pathogenesis is well established. In contrast, the composition and dynamics of the gut virome are largely unknown; particularly lacking are studies in pregnancy. We used comprehensive virome capture

Protocols

TransPlex® kits' amplification products integrate seamlessly into existing Agilent workflows for microarray target expression analyses.

TransPlex® kits' amplification products integrate seamlessly into existing Agilent workflows for microarray target expression analyses.

TransPlex® kits' amplification products integrate seamlessly into existing Agilent workflows for microarray target expression analyses.

TransPlex® kits' amplification products integrate seamlessly into existing Agilent workflows for microarray target expression analyses.

Related Content

Transplex Whole Transcriptome Amplification FAQs on topics including whole transcriptome steps, RNA source, including archival fixed tissue, library purification, quantitation of the product and downstream applications

Transplex Whole Transcriptome Amplification FAQs on topics including whole transcriptome steps, RNA source, including archival fixed tissue, library purification, quantitation of the product and downstream applications

Transplex Whole Transcriptome Amplification FAQs on topics including whole transcriptome steps, RNA source, including archival fixed tissue, library purification, quantitation of the product and downstream applications

Transplex Whole Transcriptome Amplification FAQs on topics including whole transcriptome steps, RNA source, including archival fixed tissue, library purification, quantitation of the product and downstream applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service