Skip to Content
Merck
All Photos(3)

Key Documents

WH0001591M7

Sigma-Aldrich

Monoclonal Anti-CYP24A1 antibody produced in mouse

clone 1F8, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CP24, Anti-CYP24, Anti-MGC126273, Anti-MGC126274, Anti-P450CC24, Anti-cytochrome P450, family 24, subfamily A, polypeptide 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F8, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ELISA: suitable
capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP24A1(1591)

Related Categories

General description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein initiates the degradation of 1,25-dihydroxyvitamin D3, the physiologically active form of vitamin D3, by hydroxylation of the side chain. In regulating the level of vitamin D3, this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

CYP24A1 (NP_000773, 415 a.a. ~ 514 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LMLNTQVLGSSEDNFEDSSQFRPERWLQEKEKINPFAHLPFGVGKRMCIGRRLAELQLHLALCWIVRKYDIQATDNEPVEMLHSGTLVPSRELPIAFCQR

Biochem/physiol Actions

Cytochrome P450 family 24 subfamily A member 1 (CYP24A1) plays a vital role in side-chain oxidation of steroid hormone vitamin D. Variation in or increased expression of CYP24A1 results in colorectal cancer (CRC); thus, CYP24A1 has potential as a biomarker for CRC. Biallelic mutation of CYP24A1 is observed in patients with idiopathic infantile hypercalcemia (IIH), low parathyroid hormone (PTH), renal disease, and it might also increase the risk of nephrocalcinosis in adults. Promoter polymorphism in the CYP24A1 gene leads to thechronic inflammatory skin disease called atopic dermatitis (AD) in adults. CYP24A1 transcript is highly expressed in ovary and lung tumors, but its expression is decreased in breast tumor when compared to the analogous normal tissues.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole Ball et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 38(3), 414-426 (2023-01-11)
Loss-of-function mutations in the CYP24A1 protein-coding region causing reduced 25 hydroxyvitamin D (25OHD) and 1,25 dihydroxyvitamin D (1,25(OH)2 D) catabolism have been observed in some cases of infantile hypercalcemia type 1 (HCINF1), which can manifest as nephrocalcinosis, hypercalcemia and adult-onset
M Cools et al.
Bone, 81, 89-96 (2015-06-29)
Bi-allelic CYP24A1 mutations can cause idiopathic infantile hypercalcemia (IIH), adult-onset nephrocalcinosis, and possibly bone metabolism disturbances. It is currently unclear if heterozygous carriers experience clinical problems or biochemical abnormalities. Our objective is to gain insight in the biochemical profile and
Xabier Garcia-Albeniz et al.
British journal of cancer, 114(2), 221-229 (2016-01-15)
Menopausal hormone therapy (MHT) use has been consistently associated with a decreased risk of colorectal cancer (CRC) in women. Our aim was to use a genome-wide gene-environment interaction analysis to identify genetic modifiers of CRC risk associated with use of
Hongyan Sun et al.
Human pathology, 50, 101-108 (2016-03-22)
Our study aims to fully evaluate clinicopathological and prognostic values of CYP24A1 in colorectal cancer (CRC) patients. Tissue microarrays of formalin-fixed and paraffin-embedded tumor samples and matched adjacent nontumor colorectal tissues from 99 CRC patients were studied for CYP24A1 protein
Mark G Anderson et al.
Cancer chemotherapy and pharmacology, 57(2), 234-240 (2005-09-24)
1,25-dihydroxyvitamin D3 (1,25(OH)2D3) and its analogues have been shown to inhibit proliferation of human cancer cells mediated by vitamin D receptor (VDR). The over-expression of 25-hydroxyvitamin D-24-hydroxylase (CYP24A1), an enzyme involved in the metabolism of 1,25(OH)2D3 and its analogues, is

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service