Skip to Content
Merck
All Photos(1)

Key Documents

W2270

Sigma-Aldrich

Wiskostatin

Synonym(s):

1-(3,6-dibromocarbazol-9-yl)-3-(dimethylamino)propan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18Br2N2O
CAS Number:
Molecular Weight:
426.15
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)CC(O)Cn1c2ccc(Br)cc2c3cc(Br)ccc13

InChI

1S/C17H18Br2N2O/c1-20(2)9-13(22)10-21-16-5-3-11(18)7-14(16)15-8-12(19)4-6-17(15)21/h3-8,13,22H,9-10H2,1-2H3

InChI key

XUBJEDZHBUPBKL-UHFFFAOYSA-N

General description

Wiskostatin is a dibrominated carbazole. The N-alkylated side chain has a chiral hydroxyl group and a terminal tertiary amine.

Application

Wiskostatin has been used:
  • as a small-molecule inhibitor of Wiskott-Aldrich syndrome protein (WASP) in human osteosarcoma U2OS cells and mouse-tail fibroblast cell lines
  • as a neural (N)-WASP inhibitor in cultured neurons and in the human embryonic kidney (HEK293) cells expressing HA-Parkin
  • as a neural (N)-WASP inhibitor in dorsal root ganglion (DRG) cells to investigate vascular endothelial growth factor (VEGF) effect on the actin related protein 2/3 complex (Arp 2/3)

Biochem/physiol Actions

Wiskostatin is a selective inhibitor of N-WASP, a ubiquitously expressed member of the Wiskott-Aldrich Syndrome protein (WASp) family that regulates actin polymerization. Wiskostatin inhibits actin-dependent cellular functions, including migration, transmembrane transport and phagocytosis.
Wiskostatin mediates the inhibition of cytokinesis. It interacts with WASP especially at the regulatory guanosine-5-triphosphate (GTP)ase-binding domain (GBD).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Bompard et al.
BMC cell biology, 9, 42-42 (2008-08-01)
Cytokinesis is the final step of cell division taking place at the end of mitosis during which the cytoplasmic content and replicated chromosomes of a cell are equally partitioned between the two daughter cells. This process is achieved by the
Chaohong Liu et al.
PLoS biology, 11(11), e1001704-e1001704 (2013-11-14)
Negative regulation of receptor signaling is essential for controlling cell activation and differentiation. In B-lymphocytes, the down-regulation of B-cell antigen receptor (BCR) signaling is critical for suppressing the activation of self-reactive B cells; however, the mechanism underlying the negative regulation
Alexis Cotto-Rosario et al.
Microorganisms, 10(11) (2022-11-25)
New therapeutic agents for cryptosporidiosis are a critical medical need. The marine organic compound, tartrolon E (trtE), is highly effective against multiple apicomplexan parasites, including Cryptosporidium. Understanding the mechanism of action of trtE is required to advance in the drug
R Wollman et al.
Nature cell biology, 14(12), 1261-1269 (2012-11-13)
The actin cortex both facilitates and hinders the exocytosis of secretory granules. How cells consolidate these two opposing roles was not well understood. Here we show that antigen activation of mast cells induces oscillations in Ca(2+) and PtdIns(4,5)P(2) lipid levels
Jeffrey R Peterson et al.
Nature structural & molecular biology, 11(8), 747-755 (2004-07-06)
Current drug discovery efforts focus primarily on proteins with defined enzymatic or small molecule binding sites. Autoregulatory domains represent attractive alternative targets for small molecule inhibitors because they also occur in noncatalytic proteins and because allosteric inhibitors may avoid specificity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service