Skip to Content
Merck
All Photos(3)

Key Documents

V8012

Sigma-Aldrich

Anti-V5 antibody, Mouse monoclonal

clone V5-10, purified from hybridoma cell culture

Synonym(s):

V5 Tag Monoclonal Antibody, V5 antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

V5-10, monoclonal

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 1-2 μg/mL
western blot: 0.5-1 μg/mL

shipped in

dry ice

storage temp.

−20°C

General description

Monoclonal Anti-V5 (mouse IgG1 isotype) is derived from the V5-10 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from a BALB/c mouse immunized with a synthetic peptide conjugated to KLH.
The mouse monoclonal Anti-V5 antibody reacts specifically with the V5-tagged recombinant fusion proteins expressed in transfected mammalian cells or produced by in vitro translation.
The mouse mooclonal Anti-V5 antibody reacts specifically with the V5-taged recombinant fusion proteins expressed in transfected mammalian cells or produced by in vitro translation.

Specificity

Recognizes V5 Tag (GKPIPNPLLGLDST) fusion proteins

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in "frost-free" freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Ortuno et al.
F1000Research, 5, 137-137 (2016-03-22)
A common pathological hallmark of age-related neurodegenerative diseases is the intracellular accumulation of protein aggregates such as α-synuclein in Parkinson's disease, TDP-43 in ALS, and tau in Alzheimer's disease. Enhancing intracellular clearance of aggregation-prone proteins is a plausible strategy for
Blocking Double-Stranded RNA-Activated Protein Kinase PKR by Japanese Encephalitis Virus Nonstructural Protein 2A
Tu YC, et al.
Journal of Virology, 86(19), 10347-10358 (2012)
Ernesto Ciabatti et al.
Cell, 170(2), 382-392 (2017-07-12)
Neural networks are emerging as the fundamental computational unit of the brain and it is becoming progressively clearer that network dysfunction is at the core of a number of psychiatric and neurodegenerative disorders. Yet, our ability to target specific networks
Sebastian Wissel et al.
G3 (Bethesda, Md.), 6(8), 2467-2478 (2016-06-10)
Traditional loss-of-function studies in Drosophila suffer from a number of shortcomings, including off-target effects in the case of RNA interference (RNAi) or the stochastic nature of mosaic clonal analysis. Here, we describe minimal in vivo GFP interference (miGFPi) as a
Nagarjuna Reddy Pamudurti et al.
Molecular cell, 66(1), 9-21 (2017-03-28)
Circular RNAs (circRNAs) are abundant and evolutionarily conserved RNAs of largely unknown function. Here, we show that a subset of circRNAs is translated in vivo. By performing ribosome footprinting from fly heads, we demonstrate that a group of circRNAs is associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service