Skip to Content
Merck
All Photos(1)

Key Documents

V6758

Sigma-Aldrich

VEGF Receptor-2 (Flk-1, KDR)/Fc Chimera human

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Vascular Endothelial Growth Factor Receptor‑2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in NSO cells

Assay

>90% (SDS-PAGE)

form

lyophilized powder

potency

10-40 ng/mL ED50

mol wt

predicted mol wt ~110 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... KDR(3791)

Application

VEGF Receptor-2 (Flk-1, KDR)/Fc Chimera human is a homodimeric protein contains extracellular domain of human VEGF R2 (KDR) fused to the 6x histidine tagged Fc portion of human IgG1 by the peptide (EGF). The product was used to stuyd vasculogenesis and angiogenesis and as marker for pluripotent hematopoietic stem cells.

Biochem/physiol Actions

VEGF Receptor-2 (VEGF R2) also known as Flk-1 or KDR belongs to class III subfamily of RTKs is a potent VEGF antagonist and is mainly expressed in endothelial cells. VEGF R2 can stimulate PLC-gamma pathway which in turn activate MAP kinase and subsequent strong signalling as well as can partly induces mitotic signals in NIH3T3 fibroblasts. KDR also facilitates the release of nitric oxide in endothelial cells and assist in the regulation of the vasculature.
VEGF receptors, class III receptor tyrosine kinases, have seven Ig like extracellular motifs and a tyrosine kinase intracellular domain split by a kinase insert sequence. VEGFR 1 and 2 are both expressed in an endothelial cell-specific manner.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 2.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to inhibit the VEGF-dependent proliferation of human umbilical vein endothelial cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Ziegler et al.
Science (New York, N.Y.), 285(5433), 1553-1558 (1999-09-08)
Studies on pluripotent hematopoietic stem cells (HSCs) have been hindered by lack of a positive marker, comparable to the CD34 marker of hematopoietic progenitor cells (HPCs). In human postnatal hematopoietic tissues, 0.1 to 0.5% of CD34(+) cells expressed vascular endothelial
Yanelys Morera et al.
Vaccine, 35(28), 3582-3590 (2017-05-26)
CIGB-247 is a cancer therapeutic vaccine, based on recombinant modified human vascular endothelial growth factor (VEGF) as antigen, in combination with the adjuvant VSSP, a bacterially-derived adjuvant. The vaccine have demonstrated efficacy in several murine malignancy models. These studies supported
T Takahashi et al.
Oncogene, 14(17), 2079-2089 (1997-05-01)
KDR/Flk-1 tyrosine kinase, one of the two receptors for Vascular Endothelial Growth Factor (VEGF) has been shown to generate the major part of mitotic signals in endothelial cells, although the mechanisms are poorly understood. Here we examined the processing and
J Kroll et al.
Biochemical and biophysical research communications, 265(3), 636-639 (1999-12-22)
VEGF-A induces angiogenesis and regulates endothelial function via production and release of nitric oxide (NO), which is produced by endothelial nitric oxide synthase (eNOS). While the upregulation of eNOS expression has been shown to be mediated via VEGF receptor KDR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service