Skip to Content
Merck
All Photos(2)

Key Documents

U7258

Sigma-Aldrich

Anti-Ubiquilin-1 antibody produced in rabbit

enhanced validation

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DA41, Anti-DSK2, Anti-Protein-linking IAP to cytoskeleton 1, PLIC-1, Anti-UBQLN1, Anti-XDRP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~62 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~0.5 mg/mL

technique(s)

western blot: 0.5-1.0 μg/mL using lysates of HEK-293T cells and COS7 cells expressing human ubiquilin-1.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBQLN1(29979)

General description

Ubiquilin-1 (UBQLN1) is a ubiquitin-like (UBL) protein, that contains UBL and ubiquitin-associated (UBA) domains in its N- and C-termini, respectively. UBQLN1 gene is located on the human chromosome at 9q21.32.

Specificity

Anti-Ubiquilin-1 specifically recognizes human ubiquilin-1.

Application

Anti-Ubiquilin-1 antibody produced in rabbit has been used in:
  • immunoblotting
  • immunohistochemistry
  • immunofluorescence

Biochem/physiol Actions

Ubiquilin-1 (UBQLN1) protein plays a major role in regulating the proteasomal degradation of several proteins including the presenilins PS1 and PS2. This protein is also involved in the degradation of cyclin A, γ-aminobutyric acid (GABA) receptor, and hepatitis C virus RNA-dependent RNA polymerase proteins. This protein might play a general role in neurodegenerative diseases. UBQLN1 genetic variants are associated with an increased risk of Alzheimer′s disease (AD). Downregulation of UBQLN1 gene modulates PS1 endoproteolysis. Downregulation of this gene is also involved in regulating the protein levels of nicastrin and presenilin enhancer 2 (PEN-2). It also increases the rate of β-amyloid precursor proteins (APP) maturation and trafficking through the secretory pathway associated with increased production of secretory β-amyloid precursor proteins (sAPP) and amyloid β (Aβ). UBQLN1 protein is associated with neurofibrillary tangles in AD brain and Lewy bodies in Parkinson′s disease (PD).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of ubiquilin, a novel presenilin interactor that increases presenilin protein accumulation
Mah A L, et al.
The Journal of cell biology, 151(4), 847-862 (2000)
Relationship between ubiquilin-1 and BACE1 in human Alzheimer's disease and APdE9 transgenic mouse brain and cell-based models
Natunen T, et al.
Neurobiology of Disease, 85(6), 187-205 (2016)
Gender-specific association of ATP-binding cassette transporter 1 (ABCA1) polymorphisms with the risk of late-onset Alzheimer's disease
Sundar P D, et al.
Neurobiology of Aging, 28(6), 856-862 (2007)
Katsuyoshi Mizukami et al.
Neuropathology : official journal of the Japanese Society of Neuropathology, 34(1), 11-18 (2013-07-23)
This post mortem immunohistochemical study examined the localization and distribution of ubiquilin-1 (UBL), a shuttle protein which interacts with ubiquitin and the proteasome, in the hippocampus from Alzheimer's disease (AD) dementia cases, and age-matched cases without dementia. In Braak stages
Unbiased screen reveals ubiquilin-1 and-2 highly associated with huntingtin inclusions
Rutherford N J, et al.
Brain Research, 1524(6), 62-73 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service